Dicyklohexylkarbodiimid 25% ∼1.2 mol/L pyridine

Leverantör: TCI
Fara

Synonyms: N,N'-Dicyklohexylkarbodiimid

D0437-100ML
TCIAD0437-100MLEA 1950 SEK
TCIAD0437-100ML
Dicyklohexylkarbodiimid 25% ∼1.2 mol/L pyridine
Dicyklohexylkarbodiimid
Formel: C₁₃H₂₂N₂
MW: 206.33 g/mol
Flampunkt: 113
Storage Temperature: Kylskåp
MDL Number: MFCD00011659
CAS nummer: 538-75-0
EINECS: 208-704-1
UN: 3286
ADR: 3 II
Merck Index: 14,03096
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6c3b48fa1808b6986b8dc974dc67c597885ff08&vid=575f3361e01ddde0f176b745732d2c9720bcf29c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Concentration (Nonaqueous Titration) 25.0 to 27.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR