4-Nitro-m-phenylenediamine ≥98.0%

Leverantör: TCI
Varning

Synonyms: 4-Nitro-1,3-phenylenediamine

D0104-25G
TCIAD0104-25GEA 1050 SEK
TCIAD0104-25G
4-Nitro-m-phenylenediamine ≥98.0%
4-Nitro-m-phenylenediamine
Formel: C₆H₇N₃O₂
MW: 153.14 g/mol
Smältpunkt: 159
Storage Temperature: Omgivande
MDL Number: MFCD00025289
CAS nummer: 5131-58-8
EINECS: 225-876-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72295297645463d64a8ef593cc2014ab5c1e8a6&vid=e72295297645463d64a8ef593cc2014ab5c1e8a6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 161.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR