Carbon nanotube walled_single, Length: ∼5 µm, Ø <∼3nm (>85%)

Leverantör: TCI
Varning

Synonyms: CNT

C3133-200MG
TCIAC3133-200MGEA 2425 SEK
TCIAC3133-200MG
Carbon nanotube walled_single, Length: ∼5 µm, Ø <∼3nm (>85%)
Carbon nanotube
Formel: C
MW: 12.011 g/mol
MDL Number: MFCD00133992
CAS nummer: 308068-56-6

Order Now

Specifikation testresultat

Appearance Black powder to crystal
Purity (TPO Method) min. 85 %
Diameter max. 3 nm
Length min. 5 micro_m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR