2-Ethylhexyl-2-cyano-3,3-diphenylacrylate 98,0 GC_ANALYSMETOD

Leverantör: TCI

Synonyms: 2-Ethylhexyl 2-cyano-3,3-diphenylacrylate, Octyl 2-Cyano-3,3-diphenylacrylate, 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octocrylene

C0968-25G C0968-500G C0968-100G
TCIAC0968-25GEA 375 SEK
TCIAC0968-25G TCIAC0968-500G TCIAC0968-100G
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate 98,0 GC_ANALYSMETOD
2-Ethylhexyl-2-cyano-3,3-diphenylacrylate
Formel: C₂₄H₂₇NO₂
MW: 361.49 g/mol
Kokpunkt: 218°C
Smältpunkt: –10°C
Flampunkt: 203°C
MDL Number: MFCD00059260
CAS nummer: 6197-30-4
EINECS: 228-250-8
Merck Index: 14,06756

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR