Allyl chloroacetate 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: Chloroacetic acid allyl ester, Allyl 2-chloroacetate

C0090-25G C0090-500G
TCIAC0090-25GEA 495 SEK
TCIAC0090-25G TCIAC0090-500G
Allyl chloroacetate 98,0 GC_ANALYSMETOD
Allyl chloroacetate
Formel: C₅H₇ClO₂
MW: 134.56 g/mol
Kokpunkt: 164°C
Flampunkt: 61°C
MDL Number: MFCD00058936
CAS nummer: 2916-14-5
EINECS: 220-839-8

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR