4'-Bromovalerophenone 98,0 GC_ANALYSMETOD

Leverantör: TCI

B4733-5G B4733-25G
TCIAB4733-5GEA 270 SEK
TCIAB4733-5G TCIAB4733-25G
4'-Bromovalerophenone 98,0 GC_ANALYSMETOD
4'-Bromovalerophenone
Formel: C₁₁H₁₃BrO
MW: 241.13 g/mol
Kokpunkt: 170°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000107
CAS nummer: 7295-44-5
EINECS: 230-729-1

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR