4-Butylmorpholine 98,0 GC, T

Leverantör: TCI
Varning

B4602-5ML B4602-25ML
TCIAB4602-5MLEA 855 SEK
TCIAB4602-5ML TCIAB4602-25ML
4-Butylmorpholine 98,0 GC, T
4-Butylmorpholine
Formel: C₈H₁₇NO
MW: 143.23 g/mol
Kokpunkt: 78°C
MDL Number: MFCD00023401
CAS nummer: 1005-67-0
UN: 1993
ADR: 3 III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR