(R)-3,3'-Bis[[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]-1,1'-bi-2-naphthol 92,0 HPLC_ANALYSMETOD

Leverantör: TCI

B4485-50MG
TCIAB4485-50MGEA 3425 SEK
TCIAB4485-50MG
(R)-3,3'-Bis[[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]-1,1'-bi-2-naphthol 92,0 HPLC_ANALYSMETOD
(R)-3,3'-Bis[[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]-1,1'-bi-2-naphthol
Formel: C₆₆H₅₄N₄O₂
MW: 935.18 g/mol
Storage Temperature: Frys
CAS nummer: 1620285-48-4

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 92.0 area%
Elemental analysis (Nitrogen) 5.50 to 6.30 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR