N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformimidamide

B4362-1G B4362-5G
TCIAB4362-1GEA 2825 SEK
TCIAB4362-1G TCIAB4362-5G
N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine ≥98.0% (by GC, titration analysis)
N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine
Formel: C₁₀H₁₀BrN₃
MW: 252.12 g/mol
CAS nummer: 39255-60-2

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR