2-Bromophenethylamine ≥98.0%

Leverantör: TCI
Fara

Synonyms: 2-(2-Bromophenyl)ethylamine

B3938-5G B3938-25G
TCIAB3938-5GEA 1100 SEK
TCIAB3938-5G TCIAB3938-25G
2-Bromophenethylamine ≥98.0%
2-Bromophenethylamine
Formel: C₈H₁₀BrN
MW: 200.08 g/mol
Kokpunkt: 85°C
MDL Number: MFCD01529874
CAS nummer: 65185-58-2
UN: 2735
ADR: 8 II

Order Now

Specifikation testresultat

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR