2-Bromobenzyl mercaptan 98,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

Synonyms: 2-Bromobenzylmercaptan, o-Bromotoluene-α-thiol

B3747-5G B3747-25G
TCIAB3747-5GEA 1750 SEK
TCIAB3747-5G TCIAB3747-25G
2-Bromobenzyl mercaptan 98,0 GC_ANALYSMETOD
2-Bromobenzyl mercaptan
Formel: C₇H₇BrS
MW: 203.1
Kokpunkt: 82°C
MDL Number: MFCD04038925
CAS nummer: 143888-85-1
UN: 2810
ADR: 6.1 III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR