1-Bromo-7-chloroheptane ≥95.0%

Leverantör: TCI
Varning

B3537-5G
TCIAB3537-5GEA 3200 SEK
TCIAB3537-5G
1-Bromo-7-chloroheptane ≥95.0%
1-Bromo-7-chloroheptane
Formel: C₇H₁₄BrCl
MW: 213.54 g/mol
Kokpunkt: 118°C
MDL Number: MFCD00270371
CAS nummer: 68105-93-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR