2-Bromophenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: (2-Bromophenyl)boronic acid, Dihydroxy(2-bromophenyl)borane, 2-Bromobenzeneboronic acid, (2-Bromophenyl)metaboric acid

B2889-5G
TCIAB2889-5GEA 845 SEK
TCIAB2889-5G
2-Bromophenylboronic acid (contains varying amounts of Anhydride)
2-Bromophenylboronic acid
Formel: C₆H₆BBrO₂
MW: 200.83 g/mol
MDL Number: MFCD01114672
CAS nummer: 244205-40-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cefc31502e7bce160489b0e29a0424ef89d4a8cb&vid=cefc31502e7bce160489b0e29a0424ef89d4a8cb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR