1-Bromo-2-fluorobenzene 99,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 2-Bromofluorobenzene

B0883-25G B0883-500G B0883-100G
TCIAB0883-25GEA 495 SEK
TCIAB0883-25G TCIAB0883-500G TCIAB0883-100G
1-Bromo-2-fluorobenzene 99,0 GC_ANALYSMETOD
1-Bromo-2-fluorobenzene
Formel: C₆H₄BrF
MW: 175 g/mol
Kokpunkt: 158°C
Smältpunkt: –8°C
Flampunkt: 48°C
Storage Temperature: Omgivande
MDL Number: MFCD00000282
CAS nummer: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cd08a6c0b62aeb13dd2c9071a815868310cbb97&vid=81f03af473369458c88b09b1e60f10aff9ac1348&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR