6-Bromo-2-naphthol 98,0 GC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

B0621-25G B0621-100G
TCIAB0621-25GEA 430 SEK
TCIAB0621-25G TCIAB0621-100G
6-Bromo-2-naphthol 98,0 GC_ANALYSMETOD
6-Bromo-2-naphthol
Formel: C₁₀H₇BrO
MW: 223.07 g/mol
Smältpunkt: 128
MDL Number: MFCD00004081
CAS nummer: 15231-91-1
EINECS: 239-279-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9285e654966b375d9035cbe0dd5b7a455434e8e&vid=b9285e654966b375d9035cbe0dd5b7a455434e8e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 134.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR