3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride 98,0 HPLC_ANALYSMETOD

Leverantör: TCI
Varning

Synonyms: 2-Phenyl-3-(4-nitrophenyl)-5-methyltetrazolium Chloride

B0311-100MG
TCIAB0311-100MGEA 590 SEK
TCIAB0311-100MG
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride 98,0 HPLC_ANALYSMETOD
3-(4-Nitrophenyl)-5-methyl-2-phenyltetrazolium chloride
Formel: C₁₄H₁₂ClN₅O₂
MW: 317.73 g/mol
MDL Number: MFCD00060011
UN: 1325
ADR: 4.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e502432d985d65272e6080f80693526d42e7acd&vid=ff5c86624fa50c0a15cc67cea0b3a284ccecd86b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR