N-Hydroxysuccinimide 98,0 GC, T Coupling Agent for Peptides Synthesis

Leverantör: TCI

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

B0249-25G
TCIAB0249-25GEA 645 SEK
TCIAB0249-25G
N-Hydroxysuccinimide 98,0 GC, T Coupling Agent for Peptides Synthesis
N-Hydroxysuccinimide
Formel: C₄H₅NO₃
MW: 115.09 g/mol
Kokpunkt: 363
Storage Temperature: Kylskåp
MDL Number: MFCD00005516
CAS nummer: 6066-82-6
EINECS: 228-001-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18f158051137d95734d47f7ef4e30c3686387bab&vid=a3c68ad3a3e8b4e7cdbe36d20489c5fb86ab77bb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C
Solubility in Dioxane lmost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR