3-Amino-4-methylphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

Synonyms: 3-Amino-4-methylbenzeneboronic Acid, 3-Amino-p-tolylboronic Acid, 3-Amino-4-methylphenylboronic Acid

A2745-1G A2745-5G
TCIAA2745-1GEA 1275 SEK
TCIAA2745-1G TCIAA2745-5G
3-Amino-4-methylphenylboronic acid (contains varying amounts of Anhydride)
3-Amino-4-methylphenylboronic acid
Formel: C₇H₁₀BNO₂
MW: 150.97 g/mol
MDL Number: MFCD01074640
CAS nummer: 22237-12-3

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystaline
Purity (Nonaqueous Titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR