1,9-Pyrazoloanthrone 95,0 HPLC_ANALYSMETOD

Leverantör: TCI

Synonyms: JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125

A2548-25MG
TCIAA2548-25MGEA 1600 SEK
TCIAA2548-25MG
1,9-Pyrazoloanthrone 95,0 HPLC_ANALYSMETOD
1,9-Pyrazoloanthrone
Formel: C₁₄H₈N₂O
MW: 220.231 g/mol
Smältpunkt: 282°C
Storage Temperature: Kylskåp
MDL Number: MFCD00022289
CAS nummer: 129-56-6
EINECS: 204-955-6

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 12.1 to 13.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR