2-Amino-4-chlorobenzonitrile 98,0 GC_ANALYSMETOD

Leverantör: TCI
Fara

Synonyms: 5-Chloro-2-cyanoaniline

A2483-5G A2483-25G
TCIAA2483-5GEA 565 SEK
TCIAA2483-5G TCIAA2483-25G
2-Amino-4-chlorobenzonitrile 98,0 GC_ANALYSMETOD
2-Amino-4-chlorobenzonitrile
Formel: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00035927
CAS nummer: 38487-86-4
EINECS: 253-967-8
UN: 3439
ADR: 6.1 III

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR