ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) 98,0 Titrimetrisk Biochemical Research

Leverantör: TCI
Varning

Synonyms: ABTS, Ammonium 2,2'-(hydrazine-1,2-diylidene)bis(3-ethyl-2,3-dihydrobenzo[d]thiazole-6-sulphonate), 2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulphonic acid) diammonium salt

A2166-1G
TCIAA2166-1GEA 855 SEK
TCIAA2166-1G
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) 98,0 Titrimetrisk Biochemical Research
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate])
Formel: C₁₈H₂₄N₆O₆S₄
MW: 548.68 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00010404
CAS nummer: 30931-67-0
EINECS: 250-396-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cf68fd282977196745022cdb9555e454da6cfe6&vid=8aa65947994799d410d92f9d92ae31b28145b6cc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Peroxidase detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR