6-Aminonicotinonitrile 98,0 GC, T

Leverantör: TCI
Fara

Synonyms: 6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile

A1642-25G A1642-5G
TCIAA1642-25GEA 4450 SEK
TCIAA1642-25G TCIAA1642-5G
6-Aminonicotinonitrile 98,0 GC, T
6-Aminonicotinonitrile
Formel: C₆H₅N₃
MW: 119.13 g/mol
Kokpunkt: 250°C
Smältpunkt: 164
MDL Number: MFCD00128902
CAS nummer: 4214-73-7
UN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac33844eb1d38cda9352e637a39957e765c3742&vid=2ac33844eb1d38cda9352e637a39957e765c3742&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 161.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR