Sodium-4-aminosalicylate dihydrate 98,0 HPLC, N

Leverantör: TCI

Synonyms: Sodium 4-Aminosalicylate Dihydrate, 4-Amino-2-hydroxybenzoic acid sodium salt, 4-Aminosalicylic acid sodium salt, Sodium 4-amino-2-hydroxybenzoate dihydrate

A0423-25G A0423-500G
TCIAA0423-25GEA 855 SEK
TCIAA0423-25G TCIAA0423-500G
Sodium-4-aminosalicylate dihydrate 98,0 HPLC, N
Sodium-4-aminosalicylate dihydrate
Formel: C₇H₆NNaO₃·2H₂O
MW: 211.14 g/mol (175.12 anhy)
MDL Number: MFCD00064392
CAS nummer: 6018-19-5
EINECS: 205-091-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=076b83035ff4de28cfc79c01fb598efb0b0d313f&vid=076b83035ff4de28cfc79c01fb598efb0b0d313f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 16.0 to 18.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR