96 Well Plate, RCH PSAXS
Leverantör: Tecan
606-0501RCH
TCAT606-0501RCHEA 0 SEK
TCAT606-0501RCH
96 Well Plate, RCH PSAXS
Plattor SPE plattor
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
  • Ion-exchange
  • Liquid handler compatible
A 10 µm silica micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.