Dermatome scalpel blades, sterile
Leverantör: SWANN MORTON
9943 9944 9945
SWAN9943EA 3000 SEK
SWAN9943 SWAN9944 SWAN9945
Dermatome scalpel blades, sterile
Skalpeller Skalpellblad
These Dermatome blades are fitted into a handheld instrument which resembles an electric razor. The blade oscillates and the movement back and forth will "evenly" skim off the surface layers of skin. Electric Dermatomes are preferable for cutting thinner and longer strips of skin with a more homogeneous thickness.
  • Compatible with leading brands of Dermatome knives
  • Available in three sizes: 25 mm single segment, a 50 mm two segmental and a 75 mm three segmental blade
The Dermatome range is used in the surgical removal of damaged tissue and harvestingMore Product Information