Surgical scalpels and blades, Sabre
Leverantör: SWANN MORTON
0570 0565 0563
SWAN0570EA 505 SEK
SWAN0570 SWAN0565 233-0163
Surgical scalpels and blades, Sabre
Skalpeller Skalpeller
Carbon steel blades. These blades are available separately without handles or as ready-to-use scalpels with PS handle. They are ideal for use during Podiatry and Chiropody treatments.
  • Scalpels are sterile and diposable
  • PS handles carry a graduated measure (for indication only) on the reverse side
  • Closed edge blades are ideal for 'paring' or 'scraping' skin within Podiatry and Chiropody treatments
Förpackning: Sterile scalpels are individually packaged into a transparent thermo formed tray providing improved product visibility.
Blades only are availableMore Product Information