ThermoSafe Large-Capacity Dry Ice Storage Chests
Leverantör: Cole Parmer
03721-02 03720-00 03720-10 03721-00 03721-10
STUA03721-02EA 0 SEK
STUA03721-02 STUA03720-00 STUA03720-10 STUA03721-00 STUA03721-10
ThermoSafe Large-Capacity Dry Ice Storage Chests
Kylare Kylare, bänkmodell
High capacity storage for transport of dry ice pellets and blocks.
  • Double-wall polyethylene construction filled with urethane foam insulation
  • Perfect for short or long-term storage
  • Ideal for dispensing dry ice in a laboratory environment
  • Special formulated insulation for lowest sublimation rates
Large-Capacity dry ice storage chests feature a lightweight construction with special formulated insulation for lowest sublimation rates. Chests contain extra-thick foam plastic insulation core.
Move the large-capacity storage chests easily on the aluminumMore Product Information