EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kits
Leverantör: STEMCELL Technologies
19860RF 19860
STMC19860RFEA 4639 SEK
STMC19860RF STMC19860
EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kits
Cellseparationsmedier
Immunomagnetic isolation of untouched mouse cells
  • Fast and easy-to-use
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labelled with biotinylated antibodies. labelled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet. Desired cells are untouched and poured off into a new tube.
Observera: Product Restrictions:More Product Information