EasySep™ EasyStand™ Linkable Magnet Stands with Magnets
Leverantör: STEMCELL Technologies
18131 18135 18137
STMC18131EA 15047 SEK
STMC18131 STMC18135 STMC18137
EasySep™ EasyStand™ Linkable Magnet Stands with Magnets
Magneter
Linkable stands are available in one, four, or six piece configurations. Each kit is supplied with 1, 4, or 6 EasySep™ magnets
  • Simplify the separation of one or more samples at one time
  • Magnets included
The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time. Each EasySep™ EasyStand™ can hold a single EasySep™ magnet, and a simple system links up to 6 individual EasySep™ EasyStands™ together. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes.
More Product Information