EasySep™ Release Mouse PE Positive Selection Kit
Leverantör: STEMCELL Technologies
17656
STMC17656EA 8693 SEK
STMC17656
EasySep™ Release Mouse PE Positive Selection Kit
Cellseparationsmedier
Immunomagnetic positive selection of particle-free mouse cells labelled with PE-conjugated antibodies.
  • Highly purified cells labelled with PE-conjugated antibodies isolated from mouse tissue in less than 40 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Mouse PE Positive Selection Kit is designed to isolate cells that are labelled with PE-conjugated antibodies from mouse tissue by immunomagnetic positive selection.
Desired cells are labelled with antibodies and magnetic particles, and separated without columnsMore Product Information