RosetteSep™ Human CD4+ T Cell Enrichment Cocktails
Leverantör: STEMCELL Technologies
15022 15062
STMC15022EA 2841 SEK
STMC15022 STMC15062
RosetteSep™ Human CD4+ T Cell Enrichment Cocktails
Cellseparationsmedier
Immunodensity isolation of untouched CD4+ T cells.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Isolated cells are untouched
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human CD4+ T Cell Enrichment Cocktail is designed to isolate CD4+ T cells from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-CD4+ T cells and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such asMore Product Information