AggreWell™ EB Formation Medium
Leverantör: STEMCELL Technologies
05893
STMC05893EA 2565 SEK
STMC05893
AggreWell™ EB Formation Medium
Cellodlingsmedium
Defined, serum-free medium for generation and culture of embryoid bodies using AggreWell™ plates.
AggreWell™ EB Formation Medium is a serum-free medium that supports the survival of TeSR™-cultured human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells during generation and subsequent culture of embryoid bodies (EBs).
Observera: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').