Hollow fibre filter modules, mPES membrane, MidiKros®
Leverantör: Spectrum Laboratories
D04-E003-05-N D04-E010-05-N D04-E030-05-N D04-E100-05-N D02-E100-05-N D06-E100-05-S D06-E500-10-S D06-E500-10-N D06-E500-05-S D04-E005-05-S D04-E050-05-S D04-E050-05-N D04-E030-10-S D04-E030-10-N D04-E030-05-S D04-E010-05-S D04-E070-05-N D04-E005-05-N D04-E003-05-S D04-E300-05-N D04-E500-10-S D04-E500-10-N D04-E500-05-S D04-E500-05-N D04-E300-10-S D04-E050-10-N D04-E300-05-S D04-E050-10-S D04-E100-10-S D04-E100-10-N D04-E100-05-S D04-E070-05-S D04-E300-10-N D02-E003-05-N D02-E030-05-N D02-E010-05-S D02-E010-05-N D02-E005-05-S D02-E003-05-S D02-E005-05-N D02-E030-05-S D02-E050-10-N D06-E070-05-S D02-E050-05-N D06-E300-05-N D06-E100-10-S D06-E100-10-N D06-E100-05-N D06-E300-10-N D06-E300-10-S D06-E070-05-N D06-E050-10-S D06-E050-10-N D06-E050-05-S D06-E050-05-N D06-E030-10-S D06-E300-05-S D06-E030-05-N D06-E500-05-N D02-E070-05-S D02-E300-10-N D02-E300-05-S D02-E300-05-N D02-E100-10-S D02-E100-10-N D02-E100-05-S D06-E030-10-N D02-E500-05-S D02-E070-05-N D02-E050-10-S D02-E050-05-S D02-E030-10-S D06-E003-05-N D02-E030-10-N D06-E010-05-S D06-E010-05-N D06-E005-05-S D02-E300-10-S D06-E003-05-S D02-E500-05-N D02-E500-10-S D02-E500-10-N D06-E030-05-S D06-E005-05-N
SPECD04-E003-05-NEA 10075 SEK
SPECD04-E003-05-N SPECD04-E010-05-N SPECD04-E030-05-N SPECD04-E100-05-N SPECD02-E100-05-N SPECD06-E100-05-S SPECD06-E500-10-S SPECD06-E500-10-N SPECD06-E500-05-S SPECD04-E005-05-S SPECD04-E050-05-S SPECD04-E050-05-N SPECD04-E030-10-S SPECD04-E030-10-N SPECD04-E030-05-S SPECD04-E010-05-S SPECD04-E070-05-N SPECD04-E005-05-N SPECD04-E003-05-S SPECD04-E300-05-N SPECD04-E500-10-S SPECD04-E500-10-N SPECD04-E500-05-S SPECD04-E500-05-N SPECD04-E300-10-S SPECD04-E050-10-N SPECD04-E300-05-S SPECD04-E050-10-S SPECD04-E100-10-S SPECD04-E100-10-N SPECD04-E100-05-S SPECD04-E070-05-S SPECD04-E300-10-N SPECD02-E003-05-N SPECD02-E030-05-N SPECD02-E010-05-S SPECD02-E010-05-N SPECD02-E005-05-S SPECD02-E003-05-S SPECD02-E005-05-N SPECD02-E030-05-S SPECD02-E050-10-N SPECD06-E070-05-S SPECD02-E050-05-N SPECD06-E300-05-N SPECD06-E100-10-S SPECD06-E100-10-N SPECD06-E100-05-N SPECD06-E300-10-N SPECD06-E300-10-S SPECD06-E070-05-N SPECD06-E050-10-S SPECD06-E050-10-N SPECD06-E050-05-S SPECD06-E050-05-N SPECD06-E030-10-S SPECD06-E300-05-S SPECD06-E030-05-N SPECD06-E500-05-N SPECD02-E070-05-S SPECD02-E300-10-N SPECD02-E300-05-S SPECD02-E300-05-N SPECD02-E100-10-S SPECD02-E100-10-N SPECD02-E100-05-S SPECD06-E030-10-N SPECD02-E500-05-S SPECD02-E070-05-N SPECD02-E050-10-S SPECD02-E050-05-S SPECD02-E030-10-S SPECD06-E003-05-N SPECD02-E030-10-N SPECD06-E010-05-S SPECD06-E010-05-N SPECD06-E005-05-S SPECD02-E300-10-S SPECD06-E003-05-S SPECD02-E500-05-N SPECD02-E500-10-S SPECD02-E500-10-N SPECD06-E030-05-S SPECD06-E005-05-N
Hollow fibre filter modules, mPES membrane, MidiKros®
Filter System för tangentiellt flöde
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • Filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • These filters have FLL/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultrafiltration applications. Ultra-More Product Information