ESD Steel Shelf
Leverantör: TRESTON
852283-49
SOVE852283-49EA 695 SEK
SOVE852283-49
ESD Steel Shelf
Inredning Hyllor
Epoxy powder-coated steel shelf.
  • Light gray
Due to the dual-purpose shelf brackets, the shelf can be used either as tilted 20° or in a horizontal position. The shelf has an integrated edge, which can used either in the front or back of the shelf, depending of the use of the shelf.
Ten years warranty.