Biopsy bags, SeeThru
Leverantör: Simport Scientific
M478-1 M478-2 M478-3
SIMPM478-1EA 10700 SEK
SIMPM478-1 SIMPM478-2 SIMPM478-3
Biopsy bags, SeeThru
Påsar Provpåsar
A safe and convenient way to protect your sample during processing.
  • Solvent resistant
  • Available in three sizes
  • Seams are heat-sealed
These sturdy biopsy bags are made of white nylon thin mesh (0,2 mm Ø), reducing the risk of specimen loss during processing. They provide great tissue safety and excellent fluid exchange. In order to ease specimen removal, bags can be peeled open.
Order Now
SPECIFIKATIONER
Environmentally Preferable