Tissue processing/embedding cassettes, Unisette™, M405
Leverantör: Simport Scientific
M405-3 M405-4 M405-5 M405-6 M405-7 M405-8 M405-9 M405-10 M405-11 M405-12 M405-3T M405-4T M405-5T M405-6T M405-7T M405-8T M405-9T M405-10T M405-11T M405-12T M405-2T M405-2
SIMPM405-3EA 2500 SEK
SIMPM405-3 SIMPM405-4 SIMPM405-5 SIMPM405-6 SIMPM405-7 SIMPM405-8 SIMPM405-9 SIMPM405-10 SIMPM405-11 SIMPM405-12 720-2961 720-2962 720-2963 720-2964 SIMPM405-7T SIMPM405-8T SIMPM405-9T SIMPM405-10T SIMPM405-11T SIMPM405-12T 720-2960 SIMPM405-2
Tissue processing/embedding cassettes, Unisette™, M405
Histologikassetter Histokassetter
These cassettes are moulded from a special high density polymer and keep specimens safely submerged in liquid.
  • Resistant to the chemical action of most histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • Snap latch and hinge-lock design prevent early separation of base and lid
Lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss.
Beställningsinformation: Available in non-More Product Information