Oscillating abrasion tester, model 6160, Taber®
Leverantör: Industrial Physics
TB0180
SIMETB0180EA 0 SEK
SIMETB0180
Oscillating abrasion tester, model 6160, Taber®
Utrustning för nötningstest
The oscillating abrasion tester is used to measure the resistance of a material to surface abrasion and scratching. Its primary application is for transparent materials and coatings in lenses and windows. However it can also be used to evaluate materials such as organic coatings, plastics, metals and more.
  • Test parameters can be altered and allow the operator to select the operating stroke length (6,5 to 150 mm) and speed (100 to 200 cycles per minute)
  • May be used with abrasive slurry to perform erosion tests
  • Using a custom specimen holder (not supplied),
More Product Information