Digital timer
Leverantör: Industrial Physics
DI0085
SIMEDI0085EA 735 SEK
SIMEDI0085
Digital timer
Timer Signalur, digitalt
This dual digital timer can be pre-set for both 'in position' and 'recovery' time.
Certifikat: Compliant with the ISO 2815-2003 standard.