Microscale tissue grinder kit, KIMBLE®
Leverantör: DWK Life Sciences
SP884900-0000
SCERSP884900-0000EA 19700 SEK
SCERSP884900-0000
Microscale tissue grinder kit, KIMBLE®
Homogenisatorer Homogenisatorer, för vävnad
The kit contains a variety of the finest microscale tissue grinders suitable for use on tough connective tissue and soft tissue such as brain or liver.
  • Working with micro quantities reduces heat build-up and yields greater enzyme activity
  • Samples can be handled with minimal loss; transfers can often be eliminated
  • HDPE case with die cut foam inserts that protect grinders in transit and in storage
Certifikat: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.