Polypropylenglykol, M.W. 420

Leverantör: Roth Carl

Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG

9148.2
ROTH9148.2EA 0 SEK
ROTH9148.2
Polypropylenglykol, M.W. 420
Polypropylenglykol
Formel: H(OCH₂CH₂CH₂)nOH
MW: 1025
Storage Temperature: Omgivande
MDL Number: MFCD00084446
CAS nummer: 25322-69-4
EINECS: 500-039-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR