Isobutylacetat

Leverantör: Roth Carl
Fara

Synonyms: 2-Metylpropylacetat

5092.1
ROTH5092.1EA 0 SEK
ROTH5092.1
Isobutylacetat
Isobutylacetat
Formel: C₆H₁₂O₂
MW: 116.16 g/mol
Kokpunkt: 118
Smältpunkt: –99 °C
Densitet: 0.87 g/cm³ (20 °C)
Flampunkt: 18
Storage Temperature: Omgivande
MDL Number: MFCD00008932
CAS nummer: 110-19-0
EINECS: 203-745-1
UN: 1213
ADR: 3 II
Merck Index: 13,05147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR