Glycin

Leverantör: RESEARCH ORGANICS

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

RES9582G-A103X
RESERES9582G-A103XEA 0 SEK
RESERES9582G-A103X
Glycin
Glycin
Formel: NH₂CH₂COOH
MW: 75.07 g/mol
Kokpunkt: 181
Smältpunkt: 83
Densitet: 1.64 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008131
CAS nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR