Autoclavable Bags, Biohazard
Leverantör: RATIOLAB
7003220
RATI7003220EA 7300 SEK
RATI7003220
Autoclavable Bags, Biohazard
Påsar Avfallspåsar
Autoclavable PP bags for the disposal of contaminated labware.
  • Distinctive red colour for easy identification
  • Rectangle white indicator patch field turns into dark when autoclaved
  • BIOHAZARD symbol and safety imprint in German, English and French language
  • Autoclavable at 134 °C (20 minutes)
Observera: The bag must not be completely closed when autoclaving.