2,4,6-T (2,4,6-Trichlorophenoxy acetic acid)

Leverantör: LGC Standards PROMOCHEM

Synonyms: 2-(2,4,6-Trichlorophenoxy)acetic acid

NMIAP1375
PROONMIAP1375EA 0 SEK
PROONMIAP1375
2,4,6-T (2,4,6-Trichlorophenoxy acetic acid)
2,4,6-T (2,4,6-trichlorophenoxy acetic acid)
Formel: C₈H₅Cl₃O₃
MW: 255.4825 g/mol
MDL Number: MFCD00053498
CAS nummer: 575-89-3
EINECS: 209-394-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR