Aminokapronsyra

Leverantör: LGC Standards PROMOCHEM
Varning

Synonyms: 6-Aminocaproic Acid, H-Eahx-OH, ε-Amino-n-caproic acid, 6-Aminohexanoic acid, Aminocaproic Acid

MM0896.00
PROOMM0896.00EA 0 SEK
PROOMM0896.00
Aminokapronsyra
Aminokapronsyra
Formel: C₆H₁₃NO₂
MW: 131.17 g/mol
Kokpunkt: 257
Smältpunkt: 127
Flampunkt: 207
Storage Temperature: Omgivande
MDL Number: MFCD00008238
CAS nummer: 60-32-2
EINECS: 200-469-3
Merck Index: 13,00431

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR