Oxytetracycline hydrochloride

Leverantör: LGC Standards PROMOCHEM

MM0554.00
PROOMM0554.00EA 0 SEK
PROOMM0554.00
Oxytetracycline hydrochloride
Oxytetracycline hydrochloride
Formel: C₂₂H₂₅ClN₂O₉
MW: 496.90 g/mol
MDL Number: MFCD00135815
CAS nummer: 2058-46-0
EINECS: 218-161-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR