Acetanilid

Leverantör: LGC Standards PROMOCHEM
Varning

Synonyms: N-Phenylacetamide

ICRS0201
PROOICRS0201EA 0 SEK
PROOICRS0201
Acetanilid
Acetanilid
Formel: C₈H₉NO
MW: 135.17 g/mol
Kokpunkt: 304
Smältpunkt: 115
Densitet: 1.22 g/cm³ (20 °C)
Flampunkt: 174
MDL Number: MFCD00008674
CAS nummer: 103-84-4
EINECS: 203-150-7
Merck Index: 13,00051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR