n-dodekan

Leverantör: LGC Standards PROMOCHEM
Fara

Synonyms: Dodecane

HDF02
PROOHDF02EA 0 SEK
PROOHDF02
n-dodekan
n-dodekan
Formel: H₃C(CH₂)₁₀CH₃
MW: 170.34 g/mol
Kokpunkt: 216
Smältpunkt: –9.6 °C
Densitet: 0.7487 g/cm³ (20 °C)
Flampunkt: 74
Storage Temperature: Omgivande
MDL Number: MFCD00008969
CAS nummer: 112-40-3
EINECS: 203-967-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=616d3d75a63f52fbe2a8be1681d9233be6dc0928&vid=3268fe23453ad703df2782bd3c7ba6b939ba00cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR