Reference standards, biodiesel/diesel fuel Blends
Leverantör: LGC Standards PROMOCHEM
GBDBLEND100P20 GBDBLEND10P-20 GBDBLEND10P-50 GBDBLEND15P-20 GBDBLEND20P-20 GBDBLEND25P-20 GBDBLEND-2P-20 GBDBLEND30P-20 GBDBLEND-5P-20 GBDBLEND5P-250 GBDBLEND-5P-50 GBDBLENDBLK100 GBDBLENDBLK-20
PROOGBDBLEND100P20EA 0 SEK
PROOGBDBLEND100P20 PROOGBDBLEND10P-20 PROOGBDBLEND10P-50 PROOGBDBLEND15P-20 PROOGBDBLEND20P-20 PROOGBDBLEND25P-20 PROOGBDBLEND-2P-20 PROOGBDBLEND30P-20 PROOGBDBLEND-5P-20 PROOGBDBLEND5P-250 PROOGBDBLEND-5P-50 PROOGBDBLENDBLK100 PROOGBDBLENDBLK-20
Reference standards, biodiesel/diesel fuel Blends
Standarder Petroleum Industry Standards
These standards are intended for use as calibration and reference standards for the determination of fatty acid methyl esters (FAME) biodiesel content in diesel fuel by infrared (IR) spectroscopy. They are formulated specifically for use with ASTM Method D7371 and EN 14078.