2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)

Leverantör: LGC Standards PROMOCHEM

CIL-PCB-95-C
PROOCIL-PCB-95-CEA 0 SEK
PROOCIL-PCB-95-C
2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)
2,2',3,5',6-Pentachlorobiphenyl (PCB No. 95)
Formel: C₁₂H₅Cl₅
MW: 326.44 g/mol
CAS nummer: 38379-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR